Learn More
Seracare Life Sciences Inc ANTI-HUIGG (H+L) F(AB )2 DL633
Supplier:  Seracare Life Sciences Inc 52300438
Encompass
Affinity purified polyclonal antibody, F(ab')2 fragment, to human IgG heavy and light chains (H+L) made in goat and labeled with DyLight 633. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-673-55