Learn More
ACROBiosystems ANTI-HUM IGG-COUPLED MAG BEADS
Catalog No. NC2232754
Encompass_Preferred
Anti-Human IgG-coupled Magnetic Beads (recommended for MPCLIA)(20mg 5)Detection MethodChemiluminescenceProduct TypeMagnetic Beads (Anti-Human IgG)Quantity Size10mg / 100mgPhysical Appearancelyophilized powder mixtureParticle size2.8mBeads SurfaceHydrophilicAmount of Coupled ProteinAbout 133 pmol (20 g) Anti-Human IgG / mg BeadsBinding Capacity2-5ug human antibody or Human IgG Fc tag proteins / mg beadsEmission WavelengthMeasured relative light units (RLU) at 430 nm
This item is not returnable. View return policy
Catalog No. NC2232754 Supplier ACROBiosystems Supplier No. MPCA004100MG
Anti-Human IgG-coupled Magnetic Beads (recommended for MPCLIA)(20mg 5)Detection MethodChemiluminescenceProduct TypeMagnetic Beads (Anti-Human IgG)Quantity Size10mg / 100mgPhysical Appearancelyophilized powder mixtureParticle size2.8mBeads SurfaceHydrophilicAmount of Coupled ProteinAbout 133 pmol (20 g) Anti-Human IgG / mg BeadsBinding Capacity2-5ug human antibody or Human IgG Fc tag proteins / mg beadsEmission WavelengthMeasured relative light units (RLU) at 430 nm