Learn More
Vector Laboratories Polyclonal Rabbit anti‑Human KCNH5 / Kv10.2 Antibody (aa156‑205, IHC, WB) LS‑B975, 100ul
SDP
Catalog No. NC3522272
Encompass
Kv10.2 antibody LS-B975 is an unconjugated rabbit polyclonal antibody to human Kv10.2 (KCNH5) (aa156-205). Validated for IHC and WB. Cited in 1 publication.
Catalog No. NC3522272 Supplier Vector Laboratories Supplier No. LSB975100
Kv10.2 antibody LS-B975 is an unconjugated rabbit polyclonal antibody to human Kv10.2 (KCNH5) (aa156-205). Validated for IHC and WB. Cited in 1 publication.