Learn More
BIOHIPPO Anti-Human Alpha-1-antitrypsin/AAT Z polymer specifc Antibody (2C1), 100 µg
SDP
Catalog No. 5000859081
Encompass_Preferred
Alpha-1-antitrypsin/AAT Z polymer specifc Recombinant Antibody (clone 2C1) is a recombinant Mouse IgG1, kappa antibody specific for Alpha-1-antitrypsin/AAT Z polymer specifc. It reacts with Human. Suitable for IP, IHC, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.
Catalog No. 50-008-59081 Supplier BIOHIPPO Supplier No. BHA21405088
Alpha-1-antitrypsin/AAT Z polymer specifc Recombinant Antibody (clone 2C1) is a recombinant Mouse IgG1, kappa antibody specific for Alpha-1-antitrypsin/AAT Z polymer specifc. It reacts with Human. Suitable for IP, IHC, WB. Recombinant production ensures high specificity and lot-to-lot reproducibility. Supplied as 100 ug.