Learn More
Chondrex Inc Anti-Human Alpha 5 (IV) Antibody, Clone H53
Supplier:  Chondrex Inc 7078
Encompass_Preferred
Rat IgG2a monoclonal antibody against the human non-collagenase domain of the alpha 5 chain of type IV collagen (Alpha 5 (IV)), clone H53, unconjugated. Clone H53 recognizes Alpha 5 (IV) (Epitope: IDVEF). Use for immunohistochemistry and immunoblotting. Cross-reacts to dog, human, and mouse alpha 5 (IV) NC1. Purified by Protein G chromatography. Dilute the antibody in buffer containing 2% heterologous serum, such as normal goat serum, to prevent non-specific reactions in immunohistochemistry and immunoblotting.
Catalog No. NC1929637