Learn More
Immunology Consultants anti-Human Alpha Fetoprotein Antibody (Goat) - Affinity Purified
SDP
Catalog No. 5000435496
Encompass_Preferred
This affinity‑purified goat antibody targets human Alpha Fetoprotein (AFP), a clinically relevant biomarker associated with fetal development, hepatocellular carcinoma, and germ cell tumors. The antibody provides strong specificity and low background in ELISA, Western blotting, and immunoassay development. GFT‑80A supports research in oncology, developmental biology, and biomarker validation, offering consistent, reproducible performance across serum, plasma, and purified protein samples
Catalog No. 50-004-35496 Supplier Immunology Consultants Supplier No. GFT80A
This affinity‑purified goat antibody targets human Alpha Fetoprotein (AFP), a clinically relevant biomarker associated with fetal development, hepatocellular carcinoma, and germ cell tumors. The antibody provides strong specificity and low background in ELISA, Western blotting, and immunoassay development. GFT‑80A supports research in oncology, developmental biology, and biomarker validation, offering consistent, reproducible performance across serum, plasma, and purified protein samples