Learn More
Immunology Consultants anti-Human Alpha Fetoprotein Antibody (Mouse - Clone #A13) - Monoclonal
SDP
Catalog No. 5000435453
Encompass_Preferred
Clone A13 recognizes a distinct AFP epitope, providing complementary detection to Clone A12. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MFT‑80A‑A13 is well suited for liver disease research, tumor marker studies, and diagnostic assay development. Its consistent lot‑to‑lot performance ensures reliable AFP quantification across repeated assays
Catalog No. 50-004-35453 Supplier Immunology Consultants Supplier No. MFT80AA13
Clone A13 recognizes a distinct AFP epitope, providing complementary detection to Clone A12. It supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MFT‑80A‑A13 is well suited for liver disease research, tumor marker studies, and diagnostic assay development. Its consistent lot‑to‑lot performance ensures reliable AFP quantification across repeated assays