Learn More
Immunology Consultants anti-Human Apolipoprotein C-III Antibody (Goat) - Affinity Purified
SDP
Catalog No. 5000435364
Encompass_Preferred
This affinity‑purified goat antibody targets human Apolipoprotein C‑III (ApoC3), a key regulator of triglyceride metabolism and cardiovascular risk. Designed for high‑specificity detection in ELISA, Western blotting, and lipid‑metabolism research, the antibody supports studies of dyslipidemia, metabolic disease, and biomarker profiling. Its strong affinity and low nonspecific binding ensure clean, reproducible performance across diverse sample types, including serum and purified lipoprotein fractions
Catalog No. 50-004-35364 Supplier Immunology Consultants Supplier No. GAC380A
This affinity‑purified goat antibody targets human Apolipoprotein C‑III (ApoC3), a key regulator of triglyceride metabolism and cardiovascular risk. Designed for high‑specificity detection in ELISA, Western blotting, and lipid‑metabolism research, the antibody supports studies of dyslipidemia, metabolic disease, and biomarker profiling. Its strong affinity and low nonspecific binding ensure clean, reproducible performance across diverse sample types, including serum and purified lipoprotein fractions