Learn More
Immunology Consultants Anti-Human Beta 2 Microglobulin Antibody (Mouse - Clone #501) - Monoclonal
SDP
Catalog No. 5000435336
Encompass_Preferred
This monoclonal antibody targets a distinct epitope on human Beta‑2‑Microglobulin, providing complementary detection capabilities to Clone 500. Clone 501 supports sensitive, specific measurement of β2M in ELISA, Western blotting, and immunoassay development. Its strong binding affinity and low nonspecific signal enable clear detection across serum, plasma, cell lysates, and purified protein preparations. MB2‑80A‑501 is ideal for biomarker research, MHC‑related studies, and workflows requiring precise quantification of β2M with high reproducibility and analytical clarity
Catalog No. 50-004-35336 Supplier Immunology Consultants Supplier No. MB280A501
This monoclonal antibody targets a distinct epitope on human Beta‑2‑Microglobulin, providing complementary detection capabilities to Clone 500. Clone 501 supports sensitive, specific measurement of β2M in ELISA, Western blotting, and immunoassay development. Its strong binding affinity and low nonspecific signal enable clear detection across serum, plasma, cell lysates, and purified protein preparations. MB2‑80A‑501 is ideal for biomarker research, MHC‑related studies, and workflows requiring precise quantification of β2M with high reproducibility and analytical clarity