Learn More
STEMCELL Technologies Anti-Human Beta-Tubulin III Antibody, Clone 2G10-TB3, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60092
Encompass
Mouse monoclonal IgG2a antibody against human, mouse, rat beta-tubulin III, unconjugated
Catalog No. 50-197-5509