Learn More
American Research Products Inc Anti Human C4d Antibody, Specific Detection of Complement C4d, Suitable for Frozen and Paraffin-Embedded Tissues, 250 µL
Supplier:  American Research Products Inc 125000
Encompass
This rabbit polyclonal antibody is designed for the specific detection of human complement split product C4d by immunohistochemistry (IHC). It is suitable for frozen and paraffin-embedded tissue sections, emphasizing its versatility in research settings. The antibody is affinity-purified and generated against a synthetic peptide, ensuring high specificity and sensitivity for complement activation studies. Its recommended antigen retrieval enhances its effectiveness, setting it apart in the field.
Catalog No. NC0498553