Learn More
Biolegend PE anti-human CD185 (CXCR5)
Supplier:  Biolegend 356904
Encompass_Preferred
PE anti-human CD185 (CXCR5) [J252D4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1345146