Learn More
STEMCELL Technologies Anti-Human CD117 (c-Kit) Antibody, Clone 104D2, APC, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60087AZ
Encompass
Clone 104D2 is a mouse monoclonal IgG1 antibody that reacts with human, cynomolgus, and cow CD117 (c-Kit), an ~145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells and melanocytes. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD117 (c-Kit). Binding of the 104D2 antibody does not block binding of c-Kit ligand or antibody clone SR1. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5494