Learn More
STEMCELL Technologies Anti-Human CD123 (IL-3Rα) Antibody, Clone 6H6, PerCP-Cy5.5, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60110PS.1
Encompass
Mouse monoclonal IgG1 antibody against human, rhesus, sooty mangabey CD123 (IL-3Rα), PerCP-Cy5.5-conjugated
Catalog No. 50-197-5640
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov