Learn More
Biolegend Brilliant Violet 650™ anti-human CD137 (4-1BB)
Supplier:  Biolegend 309827
Encompass_Preferred
Brilliant Violet 650™ anti-human CD137 (4-1BB) [4B4-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. NC1073980