Learn More
STEMCELL Technologies Anti-Human CD14 Antibody, Clone M5E2, APC, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60004AZ.1
Encompass
Clone M5E2 is a mouse monoclonal antibody that reacts with human, rhesus, and cynomolgus CD14, an ~53 - 55 kDa GPI-anchored transmembrane glycoprotein expressed at high levels on the surface of peripheral blood monocytes and macrophages, and at lower levels on granulocytes. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD14. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human Myeloid Positive Selection Kit (Catalog #18653) and EasySep™ Human CD14 Positive Selection Kit (Catalog #18058); partial blocking may be observed, as well as EasySep™ HLA Whole Blood CD33 Positive Selection Kit (Catalog #18287HLA), EasySep™ HLA Whole Blood Myeloid Positive Selection Kit (Catalog #18683HLA), and EasySep™ Human Buffy Coat CD14 Positive Selection Kit (Catalog #18088). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-4901