Learn More
Biolegend Brilliant Violet 785™ anti-human CD154
Supplier:  Biolegend 310842
Encompass_Preferred
Brilliant Violet 785™ anti-human CD154 [24-31]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1281855