Learn More
Fluidigm Corp ANTI-HUMAN CD16-209BI
SDP
Supplier:  Fluidigm Corp 3209002B
Encompass
CD16, also known as low-affinity IgG receptor III (FcγRIII), is a polypeptide-anchored transmembrane protein. The CD16a isoform is expressed on the surface of NK cells, activated monocytes, macrophages, and placental trophoblasts. The CD16b isoform is expressed specifically on neutrophils. CD16a and CD16b are 95% homologous and both recognized by the antibody clone 3G8.
Catalog No. NC0894880