Learn More
STEMCELL Technologies Anti-Human CD16 Antibody, Clone 3G8, Alexa Fluor 488, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60041AD.1
Encompass
Clone 3G8 is a mouse monoclonal IgG1 antibody that reacts with human, rhesus, and cynomolgus CD16 (FcγRIII or low affinity IgG receptor III), a type 1 transmembrane glycoprotein belonging to the Ig superfamily. Verified for use with flow cytometry and cell separation assays, use this antibody in applications requiring the detection of CD16. CD16/IgG interactions can result in non-specific labeling in antibody-based detection and cell separation experiments and the 3G8 antibody may be employed as a blocking antibody to reduce non-specific binding. This antibody clone has been verified for use as a CD16 (FcγRIII receptor) blocking and/or labeling antibody with EasySep™ and RosetteSep™ Human kits. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5275