Learn More
Biolegend Brilliant Violet 421™ anti-human CD162
Supplier:  Biolegend 328808
Encompass_Preferred
Brilliant Violet 421™ anti-human CD162 [KPL-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, IHC-P; Size: 100 ug
Catalog No. NC1632676