Learn More
Fluidigm Corp Anti-Human CD195/CCR5 (NP-6G4), 50 tests
SDP
Supplier:  Fluidigm Corp 3171017A
Encompass
CD195, also known as CCR5 (CC chemokine receptor type 5), is a 45 kDa protein on the surface of white blood cells involved in the immune system and acts as a receptor for chemokines. It binds to MIP-1alpha, MIP-1beta, and RANTES and is expressed on a subset of T cells and monocytes. CD195 mediates an intracellular signal that is thought to induce cell differentiation and proliferation. CCR5 has also been shown to act as a co-receptor for R5 HIV-1 cell entry.
Catalog No. NC1409250