Learn More
STEMCELL Technologies Anti-Human CD2 Antibody, Clone RPA-2.10, PE, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60007PE
Encompass
Clone RPA-2.10 is a mouse monoclonal antibody that reacts with human, rhesus, and cynomolgus CD2, an ~50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD2. CD2 expression appears early during T cell differentiation and is critically important for T cell activation and signaling, as well as lymphocyte adhesion. The primary ligand for CD2 is CD58 (LFA-3) located on antigen-presenting cells, with additional ligands comprising CD15 (SSEA-1), CD48 and CD59. Notably, the RPA-2.10 antibody blocks the mixed lymphocyte reaction. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-4938