Learn More
STEMCELL Technologies Anti-Human CD20 Antibody, Clone 2H7, PE, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60008PE.1
Encompass
Clone 2H7 is a mouse monoclonal antibody that reacts with CD20, an ~35 kDa non-glycosylated type 1 transmembrane protein in the MS4A protein family. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of human, rhesus, and cynomolgus CD20. By associating with several proteins, CD20 is involved in initiating intracellular signaling pathways that modulate the activation, proliferation, and differentiation of B cells. CD20 is thought to form multi-subunit ion channels that regulate calcium ion flux across the plasma membrane. Clone 2H7 has also been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human Whole Blood CD20 Positive Selection Kit (Catalog #18685), EasySep™ Human CD19 Positive Selection Kit II (Catalog #17854), and EasySep™ HLA Whole Blood Lymphoid Positive Selection Kit (Catalog #18684HLA). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-4949
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov