Learn More
Biolegend Alexa Fluor 647 anti-human CD209 (DC-SIGN)
Supplier:  Biolegend 330112
Encompass_Preferred
Alexa Fluor 647 anti-human CD209 (DC-SIGN) [9E9A8]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, ICC; Size: 100 tests
Catalog No. NC1352577