Learn More
Biolegend APC anti-human CD226 (DNAM-1)
Supplier:  Biolegend 338312
Encompass_Preferred
APC anti-human CD226 (DNAM-1) [11A8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC0578204