Learn More
Fluidigm Corp ANTI-HUMAN CD25 - 169TM
SDP
Supplier:  Fluidigm Corp 3169003B
Encompass
CD25, a glycoprotein known as the low-affinity interleukin 2 receptor (IL-2R), enables T cells to respond to the growth-promoting cytokine IL-2. It is expressed on regulatory and activated T cells, activated B cells, macrophages, and monocytes.
Catalog No. 50-466-968