Learn More
Biolegend PE/Cyanine7 anti-human CD27
Supplier:  Biolegend 302838
Encompass_Preferred
PE/Cyanine7 anti-human CD27 [O323]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. NC0487010