Learn More
Biolegend Brilliant Violet 785™ anti-human CD3
Supplier:  Biolegend 300472
Encompass_Preferred
Brilliant Violet 785™ anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. NC1345972