Learn More
Biolegend Alexa Fluor 488 anti-human CD303 (BDCA-2)
Supplier:  Biolegend 354230
Encompass_Preferred
Alexa Fluor 488 anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1372410