Learn More
STEMCELL Technologies Anti-Human CD32 Antibody, Clone FLI8.26, PE, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60135PE
Encompass
Clone FLI8.26 is a mouse monoclonal antibody that reacts with human and rhesus cynomolgus CD32, an ~40 kDa type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, immunomodulation, and platelet aggregation. Verified for use with ELISA, flow cytometry, and interferometry assays, use this antibody in applications requiring the detection of CD32. The FLI8.26 antibody cross-reacts with monocytes, granulocytes, and platelets. Clone FLI8.26 has been verified to bind to FcγRIIa and FcγRIIb, and reportedly binds to FcγRIIc, which shares an identical amino acid sequence to FcγRIIb in the signal peptide, extracellular and transmembrane domains. Clone FLI8.26 inhibits binding of the FcγRIIa-specific clone IV.3 antibody in flow cytometry analyses. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5711