Learn More
STEMCELL Technologies Anti-Human CD33 Antibody, Clone P67.6, PE, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60126PE.1
Encompass
Clone P67.6 is a mouse monoclonal antibody that reacts with CD33 (Siglec-3), a 67 kDa type 1 transmembrane sialoadhesin protein of the immunoglobulin superfamily. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of human CD33. CD33 is a major marker used to differentiate myeloid from lymphoid or erythroid leukemias and functions as a sialic acid-dependent cell adhesion molecule mediating protein-glycan and protein-protein interactions between cells. It also has a putative role as an inhibitory receptor during the immune response. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5700