Learn More
STEMCELL Technologies Anti-Human CD36 Antibody, Clone FA6-152, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60084
Encompass
Clone FA6-152 is a mouse monoclonal antibody that reacts with human and rat CD36, an ~88 kDa transmembrane glycoprotein that functions as a scavenger receptor, cell adhesion molecule, and signal transducer on the surface of many types of cells including platelets, monocytes, macrophages, erythrocyte precursors, endothelial and epithelial cells, and some macrophage-derived dendritic cells. Verified for use with cell separation and flow cytometry assays, use this antibody in applications requiring the detection of CD36. Binding of the FA6-152 antibody to CD36 reportedly blocks its interaction with thrombospondin, collagen, apoptotic cells, and modified LDL, and induces agglutination of fetal but not adult erythrocytes. The epitope for FA6-152 reportedly resides within an immunodominant domain of CD36 comprising amino acids 155 - 183. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5477