Learn More
STEMCELL Technologies Anti-Human CD32 Antibody, Clone IV.3, FITC, 100 Tests
SDP
Supplier:  STEMCELL Technologies 60014AZ.1
Encompass
The HIT2 antibody reacts with CD38, an ~45 kDa type II transmembrane glycoprotein expressed by many cell types, especially leukocytes. CD38 is an ectoenzyme (ADP-ribosyl hydrolase) with both cyclase and hydrolase activities, whereby it mediates lymphocyte activation, adhesion and metabolism. CD31 is a known ligand for the receptor. In humans, CD38 is expressed by the majority of hematopoietic cells at levels which vary according to the differentiation and activation status of the cells. It is highly expressed on activated T and B cells and plasma cells, and is also found at high levels in several tissues, including brain, kidney, liver, ovary, pancreas, placenta, testis, and some types of cancerous tissue. Products are for Research Use Only, Not for Diagnostic or Therapeutic use, and are not to be administered to humans.
Catalog No. 50-197-4991