Learn More
Fluidigm Corp ANTI-HUMAN CD3E (SP34-2)-170ER
SDP
Supplier:  Fluidigm Corp 3170007B
Encompass
CD3 is a transmembrane subunit of the T cell receptor (TCR) complex. It is a member of the immunoglobulin superfamily and plays a role in antigen recognition, signal transduction, and T cell activation. CD3 is expressed by T lymphocytes (thymocytes) at the highest levels on mature cell types.
Catalog No. NC0678616