Learn More
STEMCELL Technologies Anti-Human CD40 Antibody, Clone 5C3, APC, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60113AZ
Encompass
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD40, APC-conjugated
Catalog No. 50-197-5667