Learn More
STEMCELL Technologies Anti-Human CD43 Antibody, Clone CD43-10G7, PE, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60085PE.1
Encompass
Clone CD43-10G7 is a mouse monoclonal antibody that reacts with human CD43, a type I transmembrane sialoglycoprotein expressed on the surface of most human hematopoietic cells, including thymocytes, peripheral T cells, NK cells, monocytes, granulocytes, some B cells (activated B cells and an early B cell subset in bone marrow), plasma cells, and hematopoietic progenitor cells. Low expression levels are observed on platelets. CD43 exhibits different glycoforms in the range of ~95 - 135 kDa, depending on the cell type. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD43. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5483