Learn More
Biolegend FITC anti-human CD45
Supplier:  Biolegend 368508
Encompass_Preferred
FITC anti-human CD45 [2D1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1141403