Learn More
STEMCELL Technologies Anti-Human CD45RO Antibody, Clone UCHL1, Biotin, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60097BT
Encompass
Mouse monoclonal IgG2a antibody against human, chimpanzee, common marmoset CD45RO, biotin-conjugated
Catalog No. 50-197-5525
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov