Learn More
STEMCELL Technologies Anti-Human CD66b Antibody, Clone G10F5, FITC, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60086FI
Encompass
Clone G10F5 is a mouse monoclonal antibody that reacts with human and chimpanzee CD66b (previously denoted CD67), an ~95 kDa single-chain glycosylphosphatidylinositol (GPI)-linked glycoprotein expressed by neutrophilic and eosinophilic granulocytes. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of CD66b. CD66b is a member of the carcinoembryonic antigen (CEA)-like subfamily of proteins within the immunoglobulin superfamily and possesses two Ig-like C2-type domains and one Ig-like V-type domain. CD66b appears to play a role as a signaling receptor involved in cell adhesion, phagocytosis, and chemotaxis by regulating granulocyte activation and adhesion to several proteins, including CD66c, galectin-3, fibronectin, and E-selectin. Visit STEMCELL.com for more information and related products.
Catalog No. NC1210601
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov