Learn More
STEMCELL Technologies Anti-Human CD8a Antibody, Clone RPA-T8, FITC, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60022FI.1
Encompass
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD8a, FITC-conjugated
Catalog No. 50-197-5089
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov