Learn More
Immunology Consultants Monoclonal Mouse anti-Human Epididymis Protein 4 (HE4) Antibody Clone# 9F6, 1.0 mg
SDP
Catalog No. NC2734220
Encompass_Preferred
This mouse IgG2b kappa antibody is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Human Epididymis Protein 4 (HE4) as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC2734220 Supplier Immunology Consultants Supplier No. MHE480A9F6
This mouse IgG2b kappa antibody is purified using Protein A chromatography. It is produced from tissue culture media obtained at our facility in The United States. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with Human Epididymis Protein 4 (HE4) as determined by ELISA and western blotting. Optimal working dilutions should be determined experimentally by the investigator.