Learn More
Cederlane Anti-Human Fibrinogen Aff.Pur./HRPO (Paired Abs) | 5 Plates
SDP
Catalog No. NC1820447
Encompass
The materials supplier for the capture antibody are affinity purified polyclonal anti-Fg IgG for coating ELISA plates and a yellow capped tube containing 0.5ml capture IgG. For the detecting antibody peroxidase conjugated affinity purified polyclonal anti-Fg for detection of Fg and a red capped tube containing 0.5 ml of IgG-HRP conjugate are supplied.
Catalog No. NC1820447 Supplier Cederlane Supplier No. CL20067K
The materials supplier for the capture antibody are affinity purified polyclonal anti-Fg IgG for coating ELISA plates and a yellow capped tube containing 0.5ml capture IgG. For the detecting antibody peroxidase conjugated affinity purified polyclonal anti-Fg for detection of Fg and a red capped tube containing 0.5 ml of IgG-HRP conjugate are supplied.