Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Anti-Human Foxp3 Staining Set PE, eBioscience™

Catalog No. 72577640
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-577-640 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-577-640 Supplier Invitrogen™ Supplier No. 72577640
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells.

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3 cells. The PCH101 monoclonal antibody reacts with the amino terminus of human Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55 kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The PCH101 antibody crossreacts with rhesus, chimpanzee, and cynomolgus Foxp3 PCH101 recognizes a different epitope of Foxp3 than clones 236A/E7 and 150D/E4.

  • Isotype: IgG2a, kappa
  • Laser: Blue Laser
  • Excitation/Emission: 488/575 nm
  • Reported Applications: Intracellular Staining followed by Flow Cytometric Analysis

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Product Type Anti-Human Foxp3 Staining Set
Isotype IgG2a Kappa
Cell Type Foxp3
Excitation/Emission 488/575 nm
Product Line eBioscience

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.