Learn More
MP Biomedicals™ Anti-Human Hemoglobin Monoclonal Antibody from Mouse
Shop All MP Biomedicals, Inc ProductsDescription
- Anti-Human Hemoglobin Monoclonal Antibody from Mouse, in 10 mM Phosphate Buffer at pH 7.4, containing 0.9 % NaCl and 0.02% NaN3. Hemoglobin (haemoglobin, Hb) is the iron-containing oxygen-transport protein in the red blood cells of vertebrates
- Its chief physiological function is to transport oxygen from the lungs or gills to the tissues where it releases the oxygen for the cellular respiration
Specifications
Specifications
| Antigen | Human Hemoglobin |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | HB01 |
| Host Species | Mouse |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Human |
| Form | Liquid |
| Isotype | IgG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.