Learn More
Biolegend Alexa Fluor 488 anti-human HLA-G
Supplier:  Biolegend 335918
Encompass_Preferred
Alexa Fluor 488 anti-human HLA-G [87G]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1731413