Learn More
Quidel Corporation HUMAN IC3B NEO MAB BIOTIN
SDP
Supplier:  Quidel Corporation A710
Encompass
A biotin conjugated murine monoclonal antibody to a neo-epitope expressed on iC3b.
Catalog No. NC1887681