Learn More
PBL Assay Science Anti-Human Interferon Alpha (IFN-Alpha) (All Subtype) Rabbit Polyclonal Antibody (PAb) [1e5 (100,000) units/vial]
Supplier:  PBL Assay Science 311351
Encompass_Preferred
Rabbit polyclonal antibody Anti-Human Interferon Alpha (All Subtypes) is manufactured by PBL Assay Science (Catalog #31135-1) for research use only (RUO) and packaged in 1e5 (100,000) units/vial. This PAb is designed to accurately bind with and block biological activity of all human interferon alpha subtypes. It is suitable for use in direct binding ELISA applications.
Catalog No. NC1774449