Learn More
PBL Assay Science Anti-Human Interferon Alpha (IFN-Alpha) (All Subtype) Sheep Polyclonal Antibody (PAb) [1e5 (100,000) units/vial]
Supplier:  PBL Assay Science 311101
Encompass_Preferred
Sheep polyclonal antibody Anti-Human Interferon Alpha (All Subtype) is manufactured by PBL Assay Science (Catalog #31110-1) for research use only (RUO) and packaged in 1e5 (100,000) units/vial. This PAb binds to and blocks biological activity of all human IFN-Alpha subtypes. It is suitable for use in direct binding ELISA applications.
Catalog No. 50-153-8039