Learn More
PBL Assay Science Anti-Human Interferon Alpha/Beta Receptor 2 (IFNAR2) Mouse Monoclonal Antibody, Clone MMHAR-2 (MAb) [50 micrograms (ug)]
Supplier:  PBL Assay Science 213851
Encompass_Preferred
Mouse monoclonal antibody Anti-Human Interferon Alpha/Beta Receptor 2, Clone MMHAR-2 is manufactured by PBL Assay Science (Catalog #21385-1) for research use only (RUO) and packaged in 50 micrograms (ug).This MAb blocks the biological action of human Type I IFNs at IFNAR2 and is suitable for use in neutralization, flow cytometry, immunohistochemistry, and immunoprecipitation applications.
Catalog No. 50-153-8014