Learn More
PBL Assay Science Anti-Human Interferon Beta (IFN-Beta) Mouse Monoclonal Antibody, Clone MMHB-14 (MAb) [50 micrograms (ug)]
Supplier:  PBL Assay Science 214601
Encompass_Preferred
Mouse monoclonal antibody Anti-Human Interferon Beta, Clone MMHB-14 is manufactured by PBL Assay Science (Catalog #21460-1) for research use only (RUO) and packaged in 50 micrograms (ug). This MAb neutralizes human IFN-Beta and binds to human IFN-Beta with high affinity. It is suitable for use in direct binding ELISA applications.
Catalog No. 50-153-8022